Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMPR1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA511858
Description
The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. BMPRIA binds BMP4 with high-affinity in solution and is a potent BMP-4 antagonist in vitro. In adult tissues, BMPRIA is widely expressed, with the highest expression levels detected in skeletal muscle. BMPRIA is also widely expressed during embryogenesis.
Specifications
BMPR1A | |
Polyclonal | |
BMPR1A | |
BMPRIA, 10q23del, ACVRLK3, ALK3, CD292, SKR5, ALK-3, BMP type-1A receptor, BMPR-1A, activin A receptor, type II-like kinase 3, activin receptor-like kinase 3, bone morphogenetic protein receptor type-1A, serine/threonine-protein kinase receptor R5 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
12166, 657 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
P36894, P36895 | |
BMPR1A | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human BMPR1A | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction