Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMR2 Mouse anti-Human, Mouse, Rat, Monkey, Clone: 3F6, MilliporeSigma™

Catalog No. MABD171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD171MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD171MI Supplier MilliporeSigma Supplier No. MABD171
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-BMR2 Antibody, clone 3F6 is a mouse monoclonal antibody, validated for use in western blotting, Immunofluorescence and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMR2
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F6
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Gene Symbols BMR2, PPH1, BMPR3, BRK-3, T-ALK, BMPR-II, FLJ41585, FLJ76945, BMPR2
Host Species Mouse
Immunogen Purified recombinant fragment of human BMR2 expressed in E. Coli.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.