Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNC1, Mouse, Clone: 1F4, Abnova™

Catalog No. 89016323 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-323 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-323 Supplier Abnova Corporation Supplier No. H00000646M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BNC1.

The protein encoded by this gene is a zinc finger protein present in the basal cell layer of the epidermis and in hair follicles. It is also found in abundance in the germ cells of testis and ovary. This protein is thought to play a regulatory role in keratinocyte proliferation and it may also be a regulator for rRNA transcription. This gene seems to have multiple alternatively spliced transcript variants, but their full-length nature is not known yet. There seems to be evidence of multiple polyadenylation sites for this gene. [provided by RefSeq

Sequence: GKPFPEGERPCHRESVIESSGAISQTPEQATHNSERETEHTPALIMVPREVEDGGHEHYFTPGMEPQVPFSDYMELQQRLLAGGLFSALSNRGMAFPCLEDSKELEH

Specifications

Antigen BNC1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1F4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BNC1.
Formulation PBS with no preservative; pH 7.4
Gene BNC1
Gene Accession No. NM_001717
Gene Alias BNC/BSN1/HsT19447
Gene Symbols BNC1
Host Species Mouse
Immunogen BNC1 (NP_001708, 599 a.a. ∼ 705 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.