Learn More
Description
This gene is a member of the BCL2/adenovirus E1B 19kDa-interacting protein (BNIP) family. It interacts with the E1B 19kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four protein products with identical N- and C-termini. [provided by RefSeq
Specifications
Specifications
| Antigen | BNIP1 |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 1G7 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full-length recombinant BNIP1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | BNIP1 |
| Gene Accession No. | BC010959 |
| Gene Alias | NIP1/SEC20/TRG-8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.