Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bol, boule-like (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018741 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-741 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-741 Supplier Abnova Corporation Supplier No. H00066037D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BOLL protein.

This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Sequence: METESGPQTSNQMQTDSLSPSPNPVSPVPLNNPTSAPRYGTVIPNRIFVGGIDFKTNESDLRKFFSQYGSVKEVKIVNDRAGVSKGYGFVTFETQEDAQKILQEAEKLNYKDKKLNIGPAIRKQQVGIPRSSIMPAAGTMYLTTSTGYPYTYHNGVAYFHTPEVTSVPPPWPSRSVCSSPVMVAQPIYQQPAYHYQATTQYLPGQWQWSVPQPSASSAPFLYLQPSEVIYQPVEIAQDGGCVPPPLSLMETSVPEPYSDHGVQATYHQVYAPSAITMPAPVMQPEPIKTVWSIHY

Specifications

Antigen bol, boule-like (Drosophila)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BOLL protein.
Formulation PBS with no preservative; pH 7.4
Gene BOLL
Gene Accession No. NM_197970.1
Gene Symbols BOLL
Host Species Rabbit
Immunogen BOLL (NP_932074.1, 1 a.a. ∼ 295 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66037
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.