Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bone Morphogenetic Protein 4 Mouse anti-Human, Mouse, Rat, Clone: 3H2, MilliporeSigma™

Catalog No. MAB1049MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1049MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1049MI Supplier MilliporeSigma Supplier No. MAB1049
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Bone Morphogenetic Protein 4 using this Anti-Bone Morphogenetic Protein 4 Antibody, clone 3H2 validated for use in ELISA, IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bone Morphogenetic Protein 4
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3H2
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Gene Accession No. NM_001202.3; NM_130850.2; NM_130851.2
Gene Symbols BMP4, BMP2B1, BMP2B, BMP-4, DVR4, BMP-2B, ZYME
Host Species Mouse
Immunogen Recombinant mouse BMP-4 (Masuhara, 1995).
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.