Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bone Morphogenetic Protein 6 Mouse anti-Human, Rat, Clone: Morph-6.1, MilliporeSigma™

Catalog No. MAB1048MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1048MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1048MI Supplier MilliporeSigma Supplier No. MAB1048
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Bone Morphogenetic Protein 6 Antibody, clone Morph-6.1 detects level of Bone Morphogenetic Protein 6 and has been published and validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bone Morphogenetic Protein 6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Morph-6.1
Conjugate Unconjugated
Formulation Purified immunoglobulin from culture supernatant. Lyophilized from PBS containing 5mg/mL BSA and 0.09% sodium azide. Reconstitute with 0.5 mL of sterile distilled water.
Gene Accession No. NM_001718.4
Gene Symbols BMP6, VGR1, BMP-6, VGR
Host Species Mouse
Immunogen Synthetic peptide vgr 1-29C-KLH.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.