Learn More
Bone Sialoprotein II Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Bone Sialoprotein II |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Lyophilized. Resuspend in 100μL aqua bidest. Contains no preservatives. |
| Gene Symbols | IBSP; BNSP; BSP-II; BSP; SP-II |
| Host Species | Rabbit |
| Immunogen | Purified human Bone sialoprotein |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.