Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAM1 Sheep anti-Bovine, Polyclonal, R&D Systems™

Catalog No. AF6436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6436 Supplier R&D Systems Supplier No. AF6436
This item is not returnable. View return policy

Sheep Polyclonal Antibody

SPAM1 Polyclonal antibody AF6436 Specifically detects SPAM1 in Bovine samples. It is validated for Western Blotting

Specifications

Antigen SPAM1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. AAI10184
Gene Alias EC 3.2.1.35, HYA1, HYAL1, HYAL3, HYAL5, Hyal-PH20, hyaluronidase PH-20, Hyaluronoglucosaminidase PH-20, MGC26532, PH20, PH-20, SPAG15, Sperm adhesion molecule 1, sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding), Sperm surface protein PH-20
Gene Symbols SPAM1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant bovine SPAM1, Leu36-Thr497, Accession # AAI10184
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6677
Test Specificity Detects bovine SPAM1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Bovine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.