Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BPDE, Clone: 8E11, R&D Systems™

Catalog No. 4360MC100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4360MC100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4360MC100 Supplier R&D Systems Supplier No. 4360MC100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Applications:

Competitive ELISA

Specifications

Antigen BPDE
Applications ELISA
Classification Monoclonal
Clone 800000000000
Conjugate Unconjugated
Dilution Competitive ELISA
Formulation This antibody is provided from mouse ascites, provided at 1 mg/ml in PBS, 50% glycerol. with No Preservative
Host Species Mouse
Immunogen BPDE-I-G coupled to BSA
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody recognizes both free polycyclic aromatic hydrocarbons (PAHs) and DNA adducts.
Target Species Multi-species
Content And Storage Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.