Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brachyury Mouse anti-Human, Clone: 3E4.2, MilliporeSigma™

Catalog No. 04135MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-135-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-135-MI Supplier MilliporeSigma Supplier No. 04135
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Brachyury Antibody, clone 3E4.2 is validated for use in western blotting for the detection of Brachyury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brachyury
Applications Western Blot
Classification Monoclonal
Clone 3E4.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_003172
Gene Symbols MGC104817, OTTHUMP00000017588, TF, TFT
Host Species Mouse
Immunogen Synthetic human brachyury peptide, a.a. 224-435.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.