Learn More
Brain Derived Neurotrophic Factor Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Brain Derived Neurotrophic Factor |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal antiserum in buffer containing no preservatives. Lyophilized. Reconstitute with 50μL of sterile distilled water. |
| Gene Symbols | BDNF; Abrineurin; neurotrophin; MGC34632 |
| Host Species | Rabbit |
| Immunogen | A 10 amino acid peptide corresponding to the amino-terminal of mouse Brain Derived Neurotrophic Factor (BDNF) coupled to ovalbumin. |
| Purification Method | Unpurified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.