Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brain Derived Neurotrophic Factor Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1779SPMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1779SPMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1779SPMI Supplier MilliporeSigma Supplier No. AB1779SP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Brain Derived Neurotrophic Factor Antibody detects level of Brain Derived Neurotrophic Factor and has been published and validated for use in ELISA, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brain Derived Neurotrophic Factor
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoglobulin fraction lyophilized from PBS, with no preservatives. Reconstitute with 500μL of sterile distilled water (1mg/mL).
Gene Symbols BDNF; Abrineurin; neurotrophin; MGC34632
Host Species Rabbit
Immunogen Recombinant Human Brain Derived Neurotrophic Factor (BDNF).
Quantity 500 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001709.3; NM_170731.3; NM_170732.3; NM_170733.2; NM_170734.2; NM_170735.4
Target Species Human, Rat
Content And Storage Lyophilized: −20-−70°C for up to 12 months. Reconstituted: −20-−70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.