Learn More
Brain Derived Neurotrophic Factor Chicken anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Chicken Polyclonal Antibody
Supplier: MilliporeSigma AB9042
Description
Anti-Brain Derived Neurotrophic Factor Antibody, pro detects level of Brain Derived Neurotrophic Factor and has been published and validated for use in immunoprecipitation, western blot, immunohistochemistry.
Specifications
Brain Derived Neurotrophic Factor, Pro | |
Polyclonal | |
Chicken immunoglobulin. Liquid. | |
Chicken | |
100 μL | |
NM_001709.3; NM_170731.3; NM_170732.3; NM_170733.2; NM_170734.2; NM_170735.4 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
BDNF; Abrineurin; neurotrophin; MGC34632 | |
Fusion protein from mouse proBDNF containing only the prodomain region. | |
Primary | |
Human, Mouse, Rat | |
Semi-purified |