Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCA1 Mouse anti-Human, Mouse, Clone: BR64, MilliporeSigma™

Catalog No. MAB4132MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4132MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4132MI Supplier MilliporeSigma Supplier No. MAB4132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-BRCA1 Antibody, clone BR64 is a high quality Mouse Monoclonal Antibody for the detection of BRCA1 and has been validated in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BR64
Conjugate Unconjugated
Formulation Liquid in phosphate buffer, containing azide as a preservative.
Gene Accession No. NM_007294.2; NM_007295.2; NM_007296.2; NM_007297.2; NM_007298.2; NM_007299.2; NM_007300.2; NM_007302.2; NM_007303.2; NM_007304.2; NM_007305.2
Gene Symbols BRCA1, RNF53, BRCAI, PSCP, BRCC1, IRIS
Host Species Mouse
Immunogen Purified polypeptide representing a 100-amino acid segment of the amino terminus of the BRCA1 gene product.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.