Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCA1 Mouse anti-Human, Clone: MS110, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC199
Description
Anti-BRCA1 Antibody, clone MS110 is an antibody against BRCA1 for use in Western Blotting, immunocytochemistry.
Specifications
BRCA1 | |
Monoclonal | |
Unconjugated | |
NP_009225 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
MS110 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
BRCA1, RNF53 | |
Recombinant protein corresponding to the N-terminus of human BRCA1. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction