Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-BRCA1 Clone: MS13,

Catalog No. OP93100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP93100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP93100UG Supplier MilliporeSigma™ Supplier No. OP93100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects BRCA1 Clone: MS13 in Human samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation

Inactivation of the BRCA1 gene, located at 17q21, is responsible for some forms of familial breast and ovarian cancer and sporadic ovarian cancer. BRCA1 encodes a protein of 1863 amino acids with an amino-terminal domain that exhibits similarity to the zinc-ring domain found in several regulatory and transcription factors. Mutations that disrupt this domain are some of the most frequently occurring ones linked to breast and ovarian cancer. Inhibition of BRCA1 expression leads to accelerated growth of mammary cells in vitro while loss of expression is observed during progression from benign disease to metastatic breast cancer suggesting that BRCA1 serves as a negative growth regulator. The ∽220kDa BRCA1 protein is expressed predominantly in the nucleus and appears to undergo cell cycle regulation with mRNA expression induced prior to DNA synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone MS13
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen recombinant, human BRCA1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽220kDa BRCA1 protein in MCF-7 cells. Does not react with blood group antigens or growth factor receptors such as EGFR. Sold under license of U.S. Patent 5,753,441 and 6,162,897.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.