Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRCA1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07434
Description
Detect BRCA1 with Anti-BRCA1 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in immunohistochemistry, immunoprecipitation and western blotting. This BRCA1 antibody has been tested in human samples.
Specifications
| BRCA1 | |
| Polyclonal | |
| Serum containing 0.035% sodium azide and 30% glycerol. Liquid at −20°C. | |
| Rabbit | |
| Unpurified | |
| Primary | |
| Human | |
| Serum |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| BRCA1; RNF53; BRCAI; PSCP; BRCC1; IRIS | |
| GST-tagged fusion protein corresponding to amino acids 1301-1863 of human BRCA1. | |
| 100 μL | |
| NM_007294 | |
| −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction