Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-BRCA2 Clone: 2B,

Catalog No. OP95100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP95100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP95100UG Supplier MilliporeSigma™ Supplier No. OP95100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects BRCA2 Clone: 2B in Human samples, and it is validated for Immunoprecipitation

Inherited mutations in the BRCA2 gene enhances the risk of familial breast cancer with the neoplasia related to loss of the wild type allele. Human BRCA2 encodes a 3418 amino acid protein with little homology to any known proteins. Transcription of BRCA2 appears to be co-ordinately regulated with BRCA1 during proliferation and differentiation with low level expression in early G1 and maximal expression occurring during the G1/S transition of the cell cycle. While loss of BRCA2 is associated with early onset breast cancer, the gene is also critical for embryogenesis. Although some mice nullizygous for BRCA2 die during embryogenesis, there have been BRCA2 knock-out mice that do survive to adulthood, albeit with various defects particularly in DNA repair capability. Loss of BRCA2 expression in embryogenesis does not appear to affect apoptosis, per se, but proliferation is impaired, perhaps due to enhanced expression of p21WAF1. Although the precise role for BRCA2 is still to be determined, recent studies implicate BRCA2 in DNA repair and recombination processes due to association with p53 and Rad51 and co-localization with BRCA1. BRCA2 levels, as well as BRCA1 levels, decline following DNA damage and the decrease in BRCA2 accompanies activation of p53. Interestingly, BRCA2 appears capable of specifically inhibiting p53 transcriptional activity, which implicates BRCA2 as a mediator of cell cycle control and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA2
Applications Immunoprecipitation
Classification Monoclonal
Clone 2B
Conjugate Unconjugated
Formulation In PBS.
Host Species Mouse
Immunogen a recombinant protein consisting of amino acids 1651-1821 of human BRCA2 fused to GST
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognzies the ∽460kDa BRCA2 protein in cells expressing a wild type BRCA2 gene. Sold under license of U.S. Patent 5,753,441 and 6,162,897.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.