Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-BRCA2, Polyclonal

Catalog No. CA1033100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CA1033100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CA1033100UG Supplier MilliporeSigma™ Supplier No. CA1033100UG

Rabbit Polyclonal Antibody

Specifically detects BRCA2 Clone: in Human samples, and it is validated for Immunohistochemistry (Paraffin), Immunoprecipitation

Inherited mutations in the BRCA2 gene enhances the risk of familial breast cancer with the neoplasia related to loss of the wild type allele. Human BRCA2 encodes a 3418 amino acid protein with little homology to any known proteins. Transcription of BRCA2 appears to be generally co-ordinately regulated and co-locally expressed with BRCA1 during proliferation and differentiation with low level expression in early G1 and maximal expression occurring during the G1/S transition of the cell cycle. However, BRCA1 and BRCA2 expression is differentially regulated during the development of specific endocrine tissues suggesting a gene-specific response to sex hormones. While loss of BRCA2 is associated with early onset breast cancer, the gene is critical for embryogenesis since nullizygous mice with a deletion of the 5' region of exon 11 arrest development by day 8.5 of gestation. Recently, however, mice carrying homozygous inactivating mutations due to deletion of exon 11 sequences or insertion of selectable markers in the 3' portion of the cDNA have been obtained, albeit with severe growth characteristics including lack of germinal cells, small stature, and skeletal deformities. Loss of BRCA2 expression in embryogenesis does not appear to be due to apoptosis, but proliferation is impaired, perhaps due to enhanced expression of p21WAF1. Although the precise role for BRCA2 is still to be determined, recent studies implicate BRCA2 in DNA repair and recombination processes partly due to association with the Rad51 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCA2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS.
Host Species Rabbit
Immunogen a recombinant protein consisting of amino acids 3245-3418 of human BRCA2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽400kDa BRCA2 protein in MCF-7 and HeLa cells and breast carcinoma.
Target Species Human
Content And Storage −20°C
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.