Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BRCC2, Polyclonal, Abnova

Catalog No. 89008770 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-770 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-770 Supplier Abnova Corporation Supplier No. H00414899B01

Mouse Polyclonal

Detection Antibody, Immunofluorescence, Western Blotting

Specifications

Antigen BH3-like motif containing, cell death inducer
Classification Polyclonal
Conjugate Unconjugated
Host Species Mouse
Immunogen BRCC2 (NP_001001786.1, 1 a.a. ∽108 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Target Species Human
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.