Learn More
BRD4, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human BRD4 peptide using ARM Technology.
Supplier: Abnova Corporation H00023476K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is homologous to the murine protein MCAP, which associates with chromosomes during mitosis, and to the human RING3 protein, a serine/threonine kinase. Each of these proteins contains two bromodomains, a conserved sequence motif which may be involved in chromatin targeting. This gene has been implicated as the chromosome 19 target of translocation t(15;19)(q13;p13.1), which defines an upper respiratory tract carcinoma in young people. Two alternatively spliced transcript variants have been described. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| BRD4 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| BRD4 | |
| BRD4 | |
| A synthetic peptide of human BRD4 | |
| RUO | |
| 23476 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| bromodomain containing 4 | |
| BRD4 | |
| CAP, HUNK1, HUNKI, MCAP | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.