Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Mouse, Brilliant Violet 510, Clone: 3D4, BD

Catalog No. BDB563445 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563445 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563445 Supplier BD Biosciences Supplier No. 563445
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bromodeoxyuridine (BrdU) is an analog of thymidine that can be incorporated into newly synthesized DNA by cells entering and progressing thru the DNA synthesis (S) phase of the cell cycle. The amount of BrdU that gets incorporated is dependent upon the amount of time that the cells are exposed to BrdU (pulse time), the rate of cell division, and whether the cells are in early, mid, or late S phase. Detection of incorporated BrdU allows the investigator to identify cycling cells in an asynchronous cell population and to determine cell cycle kinetics.

The 3D4 monoclonal antibody reacts with BrdU, but not other nucleotides, in single-stranded DNA. Random cleavage (nicking) of cellular DNA with DNase I permits the binding of the antibody to incorporated BrdU.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Flow Cytometry
Classification Monoclonal
Clone 3D4
Conjugate Brilliant Violet 510
Description 5-bromo-2'-deoxyuridine, 5-Bromouracil deoxyriboside, BUdR
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.