Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Mouse anti-All, Clone: AH4H7-1 / 131-14871, MilliporeSigma™

Catalog No. MAB3424MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3424MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3424MI Supplier MilliporeSigma Supplier No. MAB3424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-BrdU Antibody, clone AH4H7-1 / 131-14871 (Mouse Monoclonal Antibody) validated in flow cytometry, immunocytochemistry, immunohistochemistry to detect Bromodeoxyuridine also known as BrdU.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone AH4H7-1 / 131-14871
Conjugate Unconjugated
Formulation Liquid in 10mM Phosphate buffer, pH 7.4 containing 150mM NaCl and 0.1% sodium azide
Host Species Mouse
Immunogen Bromodeoxyuridine-bovine serum albumin concentrate
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.