Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Mouse anti-All, Clone: BU-1, MilliporeSigma™

Catalog No. 05633MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-633-MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-633-MI Supplier MilliporeSigma Supplier No. 05633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect BrdU also known as Bromodeoxyuridine with Anti-BrdU Antibody, clone BU-1 (Mouse Monoclonal Antibody), that has been demonstrated to work in immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Immunocytochemistry
Classification Monoclonal
Clone BU-1
Conjugate Unconjugated
Formulation 4 vials, each vial containing mouse culture supernatant lyophilized from 2.5 mL. The supernatant contains mycoplasma, which contribute enzymes that facilitate antibody binding. Lyophilized powder.
Host Species Mouse
Immunogen 5-iodouridine:ovalbumin conjugate
Purification Method Concentrated culture supernatant
Quantity 1 Unit
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C for 24 months.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.