Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Mouse anti-All, Clone: BU-1, MilliporeSigma™

Catalog No. MAB3510MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3510MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3510MI Supplier MilliporeSigma Supplier No. MAB3510
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use mouse monoclonal Anti-BrdU Antibody, clone BU-1, validated in Flow Cytometry, Immunocytochemistry, and Immunohistochemistry for the detection of bromodeoxyuridine (BrdU).
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone BU-1
Conjugate Unconjugated
Formulation UnPurified mouse tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL. Possible mycoplasma risk contained within thi
Host Species Mouse
Immunogen Bromodeoxyuridine
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.