Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Mouse anti-All, FITC, Clone: PRB-1, MilliporeSigma™

Catalog No. MAB3262FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
75 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3262FMI 75 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3262FMI Supplier MilliporeSigma Supplier No. MAB3262F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-BrdU Antibody, clone PRB-1, FITC conjugated is a Mouse Monoclonal Antibody for detection of Bromodeoxyuridine also known as BrdU and has been validated in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU
Applications Flow Cytometry
Classification Monoclonal
Clone PRB-1
Conjugate FITC
Formulation Purified immunoglobulin conjugated to fluorescein. Liquid in PBS with 0.1% sodium azide.
Host Species Mouse
Immunogen Iodouridine conjugated to ovalbumin.
Quantity 75 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.