Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-BRE1-A (RNF20) Clone: 8A4.2,

Catalog No. MABE948MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE948MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE948MI Supplier MilliporeSigma™ Supplier No. MABE948
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects BRE1-A (RNF20) Clone: 8A4.2 in Human samples, and it is validated for Western Blotting

E3 ubiquitin-protein ligase BRE1A (UniProt Q5VTR2; also known as BRE1 E3 ubiquitin ligase homolog, BRE1-A, hBRE1, Homolog of S. cerevisiae BRE1, RING finger protein 20) is encoded by the RNF20 (also known as BRE1, BRE1A) gene (Gene ID 56254) in human. BRE1A (RNF20) is a RING finger domain-containing protein originally identified as the E3 uqibiquitin ligase that catalyzes histone H2B Lys120 monoubiquitination in yeast. The histone chaperone FACT is reported to mediate RNF20 recruitment for chromatin remodeling to initiate homologous recombination repair (HRR). RNF20 is also shown to negatively regulate hepatic fatty acid metabolism by targeting SREBP1c for ubiquitination and subsequent proteasomal degradation. In addition, RNF20 is shown to mediated the polyubiquitination of ErbB3-binding protein 1 (Ebp1) and Transcription factor activator protein 2α (AP-2α). AP-2α functions as a negative regulator of adipogenesis by suppressing the transcription of CCAAT/enhancer binding protein (C/EBPα). Overexpression of RNF20 in 3T3-L1 cells results in a down-regulation of endogenous AP-2α and a concomitant transcriptional upregulation of C/EBPα.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRE1-A (RNF20)
Applications Western Blot
Classification Monoclonal
Clone 8A4.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q5VTR2
Gene Symbols RNF20, BRE1, BRE1A
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the third coiled-coil region of human BRE1-A (RNF20).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_062538
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.