Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brevican Mouse, Unlabeled, Clone: 2, BD

Catalog No. BDB610894 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610894 50 μg
BDB610895 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610894 Supplier BD Biosciences Supplier No. 610894
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neural tissue expresses a variety of cell surface heparan sulfate and extracellular chondroitin sulfate containing proteoglycans. Brevican, a chondroitin sulfate proteoglycan, is a member of the lectican family. Other members of this family are aggrecan, versican, and neurocan. These proteins are either heparan sulfate or keratan sulfate containing glycosylphosphatidylinositol (GPI) anchored or membrane spanning proteoglycans, or soluble chondroitin sulfate containing proteoglycans. Brevican exists as either a soluble or GPI-anchored plasma membrane protein. Brevican contains the shortest core protein of any of the family members. In addition, it is characterized by chondroitin sulfate chains, an N-terminal hyaluronic acid-binding domain (HA), an epidermal growth factor-like (EGF) repeat, and lectin-like and complement regulatory protein-like domains in its C-terminal region. Primary cerebellar astrocytes are the major site for mRNA expression and little, if any, is found in neurons. Although there is an 80 kDa N-terminal truncated form, full length brevican can be up to 145 kDa. Brevican is thought to play a role in the terminal differentiation and regulation of the adult nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brevican
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Brevican aa. 232-394
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.