Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brk Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09143MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-143-MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-143-MI Supplier MilliporeSigma Supplier No. 09143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Brk using this Anti-Brk Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brk
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 50 μg of affinity purified rabbit polyclonal IgG in 0.02M PBS, 250mM sodium chloride containing 0.1% sodium azide.
Gene Symbols PTK6; BRK; FLJ42088
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide corresponding to the N-terminal region of human Brk.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005975.2
Target Species Human
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.