Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brm Mouse, Unlabeled, Clone: 24, BD

Catalog No. BDB610390 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610390 150 μg
BDB610389 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610390 Supplier BD Biosciences Supplier No. 610390
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Several of the SNF and SWI genes of Saccharomyces cerevisiae encode proteins that are involved in the regulation of transcriptional activation. One of these proteins, SNF2/SWI2, has both . The human Brm protein has been reported to be a 180 kDa nuclear factor that acts as a transcriptional activator when fused to a heterologus DNA binding domain. Transfected Brm, expressed in cells lacking endogenous protein, can cooperate with the glucocorticoid receptor (GR) in transcriptional activation. The cooperation between Brm and GR requires the DNA binding domain of GR and the helicase domain and the P/Q-charged domain of Brm. However, Brm does not affect on several other transcription factors. The retinoblastoma protein, Rb, stimulates the transcription of a number of genes. Like Brm, Rb up-regulates glucocorticoid-receptor-mediated transcription. Brm and Rb interact in vitro and in vivo , requiring the Rb-pocket domain and the consensus Rb-binding motif of Brm.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Brm
Applications Western Blot
Classification Monoclonal
Clone 24
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.