Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brn-3a Mouse anti-Chicken, Monkey, Rat, Clone: 5A3.2, MilliporeSigma™

Catalog No. MAB1585MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1585MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1585MI Supplier MilliporeSigma Supplier No. MAB1585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Brn-3a Antibody, POU-domain protein, clone 5A3.2 detects level of Brn-3a and has been published and validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Brn-3a
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5A3.2
Conjugate Unconjugated
Formulation UnPurified mouse tissue culture supernatant from a perfusion system, filtered through a 0.2μ micron membrane prior to vialing. Product contains 20%FBS and Ciprofloxacin at final concentration of 10μg/mL.
Gene Accession No. NM_006237.3
Gene Symbols POU4F1, FLJ13449, Brn-3A, RDC-1, Oct-T1, BRN3A, RDC1
Host Species Mouse
Immunogen Amino acids 186-224 of Brn-3a fused to the T7 gene 10 protein (Xiang et al., 1995).
Purification Method Unpurified
Quantity 1 mL
Primary or Secondary Primary
Target Species Chicken, Monkey, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.