Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BSCL2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002989 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-989 Supplier Abnova Corporation Supplier No. H00026580D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BSCL2 protein.

This gene encodes protein seipin, which is located in the endoplasmic reticulum and may be important for lipid droplet morphology. Mutations in this gene have been associated with congenital generalized lipodystrophy type 2 or Berardinelli-Seip syndrome, a rare autosomal recessive disease characterized by a near absence of adipose tissue and severe insulin resistance. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MVNDPPVPALLWAQEVGQVLAGRARRLLLQFGVLFCTILLLLWVSVFLYGSFYYSYMPTVSHLSPVHFYYRTDCDSSTTSLCSFPVANVSLTKGGRDRVLMYGQPYRVTLELELPESPVNQDLGMFLVTISCYTRGGRIISTSSRSVMLHYRSDLLQMLDTLVFSSLLLFGFAEQKQLLEVELYADYRENSYVPTTGAIIEIHSKRIQLYGAYLRIHAHFTGLRYLLYNFPMTCAFIGVASNFTFLSVIVLFSYMQWVWGGIWPRHRFSLQVNIRKRDNSRKEVQRRISAHQPGAGPEGQEESTPQSDVTEDGESPEDPSGTEGQLSEEEKPDQQPLSGEEELEPEASDGSGSWEDAALLTEANLPAPAPASASAPVLETLGSSEPAGGALRQRPTCSSS

Specifications

Antigen BSCL2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BSCL2 protein.
Formulation PBS with no preservative; pH 7.4
Gene BSCL2
Gene Accession No. BC009866.2
Gene Alias GNG3LG/HMN5/MGC4694/SPG17
Gene Symbols BSCL2
Host Species Rabbit
Immunogen BSCL2 (AAH09866.2, 1 a.a. ∼ 400 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26580
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.