Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTB (POZ) domain containing 1, Mouse, Clone: 3E11, Abnova™

Catalog No. 89014313 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-313 Supplier Abnova Corporation Supplier No. H00053339M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BTBD1.

The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: EYEKKQTLGQNDTGFSCDGTANTFRVMFKEPIEILPNVCYTACATLKGPDSHYGTKGLKKVVHETPAASKTVFFFFSSPGNNNGTSIEDGQIPEIIFYT

Specifications

Antigen BTB (POZ) domain containing 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BTBD1.
Formulation PBS with no preservative; pH 7.4
Gene BTBD1
Gene Accession No. NM_025238
Gene Alias C15orf1/NS5ATP8
Gene Symbols BTBD1
Host Species Mouse
Immunogen BTBD1 (NP_079514, 384 a.a. ∼ 482 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53339
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.