Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

biotinidase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018055 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-055 Supplier Abnova Corporation Supplier No. H00000686D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BTD protein.

Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency. [provided by RefSeq

Sequence: MAHAHIQGGRRAKSRFVVCIMSGARSKLALFLCGCYVVALGAHTGEESVADHHEAEYYVAAVYEHPSILSLNPLALISRQEALELMNQNLDIYEQQVMTAAQKDVQIIVFPEDGIHGFNFTRTSIYPFLDFMPSPQVVRWNPCLEPHRFNDTEVLQRLSCMAIRGDMFLVANLGTKEPCHSSDPRCPKDGRYQFNTNVVFSNNGTLVDRYRKHNLYFEAAFDVPLKVDLITFDTPFAGRFGIFTCFDILFFDPAIRVLRDYKVKHVVYPTAWMNQLPLLAAIEIQKAFAVAFGINVLAANVHHPVLGMTGSGIHTPLESFWYHDMENPKSHLIIAQVAKNPVGLIGAENATGETDPSHSKFLKILSGDPYCEKDAQEVHCDEATKWNVNAPPTFHSEMMYDNFTLVPVWGKEGYLHVCSNGLCCYLLYERPTLSKELYALGVFDGLHTVHGTYYIQVCALVRCGGLGFDTCGQEITEATGIFEFHLWGNFSTSYIFPLFLTSGMTLEVPDQLGWENDHYFLRKSRLSSGLVTAALYGRLYERD

Specifications

Antigen biotinidase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BTD protein.
Formulation PBS with no preservative; pH 7.4
Gene BTD
Gene Accession No. NM_000060.2
Gene Symbols BTD
Host Species Rabbit
Immunogen BTD (NP_000051.1, 1 a.a. ∼ 543 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 686
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.