Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-cell translocation gene 4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017099 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-099 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-099 Supplier Abnova Corporation Supplier No. H00054766B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human BTG4 protein.

The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein can induce G1 arrest in the cell cycle. [provided by RefSeq

Sequence: MRDEIATTVFFVTRLVKKHDKLSKQQIEDFAEKLMTILFETYRSHWHSDCPSKGQAFRCIRINNNQNKDPILERACVESNVDFSHLGLPKEMTIWVDPFEVCCRYGEKNHPFTVASFKGRWEEWELYQQISYAVSRASSDVSSGTSCDEESCSKEPRVIPKVSNPKSIYQVKSVPVLFYTFFLSNSKKNALIMKTKKQKNMERTKL

Specifications

Antigen B-cell translocation gene 4
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human BTG4 protein.
Formulation PBS with no preservative; pH 7.4
Gene BTG4
Gene Accession No. BC031045
Gene Alias MGC33003/PC3B
Gene Symbols BTG4
Host Species Mouse
Immunogen BTG4 (AAH31045.1, 1 a.a. ∼ 206 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 54766
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.