Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Btk Mouse anti-Human, Unlabeled, Clone: 53/BTK, BD

Catalog No. BDB611117 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611117 150 μg
BDB611116 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611117 Supplier BD Biosciences Supplier No. 611117
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bruton's tyrosine kinase (Btk) is a nonreceptor tyrosine kinase whose function is critical for proper B cell development and signaling. It is a member of the Tec family of kinases which includes Tec and Itk. In addition to an N-terminal pleckstrin homology (PH) domain, the Tec proteins contain Src homology domains 2 and 3 (SH2 and SH3) and a stretch of 60-80 amino acids between the PH and SH3 domains termed the Tec homology domain. The activity of Btk is regulated by Src-mediated phosphorylation of the kinase domain at tyrosine 551. This event induces Btk kinase activity and subsequent autophosphorylation at tyrosine 223 in the SH3 domain. Phosphorylated Btk then associates with the cell membrane via the interaction of the PH domain with phosphatidylinositol 3, 4, 5-triphosphate. The PH domain is essential for proper activation and function of Btk. A mutation in the PH domain results in Xid, murine X-linked immunodeficiency, and human X-linked agammaglobulinemia.

Host Species: Mouse
Clone: 53
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human N-Terminal Btk aa. 2-172 Recombinant Protein
Formula Weight [Chemical]: 77kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Btk
Applications Western Blot
Classification Monoclonal
Clone 53/BTK
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human N-Terminal Btk aa. 2-172 Recombinant Protein
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.