Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Btk Mouse anti-Human, PE, Clone: 53/BTK, BD

Catalog No. BDB558527 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558527 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558527 Supplier BD Biosciences Supplier No. 558527
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bruton's tyrosine kinase (Btk) is a nonreceptor tyrosine kinase whose function is critical for proper B cell development and signaling. It is a member of the Tec family of kinases which includes Tec and Itk. This family is similar to the src family of tyrosine kinases. However, Tec family members lack the N-terminal myristylation site and the regulatory C-terminal tyrosine that are found in src proteins. In addition to an N-terminal pleckstrin homology (PH) domain, the Tec proteins contain Src homology domains 2 and 3 (SH2 and SH3) and a stretch of 60-80 amino acids between the PH and SH3 domains termed the Tec homology domain. The activity of Btk is regulated by Src-mediated phosphorylation of the kinase domain at tyrosine 551. This event induces Btk kinase activity and subsequent autophosphorylation at tyrosine 223 in the SH3 domain. Phosphorylated Btk then associates with the cell membrane via the interaction of the PH domain with phosphatidylinositol 3, 4, 5-triphosphate. The PH domain is essential for proper activation and function of Btk. A mutation in the PH domain results in Xid, murine X-linked immunodeficiency, and human X-linked agammaglobulinemia.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Btk
Applications Flow Cytometry
Classification Monoclonal
Clone 53/BTK
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human N-terminal Btk Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.