Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Btk (pY551) and Itk (pY511) Mouse, Unlabeled, Clone: 24A/BTK (Y551), BD

Catalog No. BDB558034 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558034 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558034 Supplier BD Biosciences Supplier No. 558034
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B ruton's t yrosine k inase (Btk) is a nonreceptor tyrosine kinase whose function is critical for proper B cell development and signaling. The activity of Btk is regulated by Src mediated phosphorylation of the kinase domain at tyrosine 551 (Y551). This event induces Btk kinase activity and subsequent autophosphorylation at Y223. Phosphorylated Btk then associates with the cell membrane via the interaction of the PH domain with phosphatidylinositol 3, 4, 5-triphosphate. γ1 (PLC γ1), a key regulator of calcium mobilization and extracellular signal-regulated kinase (ERK) activation. Itk is regulated by an activating transphosphorylation event in which Y511 in the kinase domain is phosphorylated by Lck.

The 24a/BTK (Y551) monoclonal antibody recognizes the Y551-phosphorylated form of human Btk and the Y511 phosphorylated form of human Itk.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Btk (pY551) and Itk (pY511)
Applications Western Blot
Classification Monoclonal
Clone 24A/BTK (Y551)
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ATK; BPK; AGMX1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human Btk Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.