Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BTRC Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535278
Description
This antibody is predicted to react with mouse based on sequence homology.
Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Recognizes and binds to phosphorylated target proteins. SCF(BTRC) mediates the ubiquitination of CTNNB1 and participates in Wnt signaling. SCF(BTRC) mediates the ubiquitination of NFKBIA, NFKBIB and NFKBIE; the degradation frees the associated NFKB1 to translocate into the nucleus and to activate transcription. Ubiquitination of NFKBIA occurs at ′Lys-21′ and ′Lys-22′. SCF(BTRC) mediates the ubiquitination of phosphorylated NFKB1/nuclear factor NF-kappa-B p105 subunit, ATF4, SMAD3, SMAD4, CDC25A, DLG1, FBXO5 and probably NFKB2. SCF(BTRC) mediates the ubiquitination of phosphorylated SNAI1. May be involved in ubiquitination and subsequent proteasomal degradation through a DBB1-CUL4 E3 ubiquitin-protein ligase. Required for activation of NFKB-mediated transcription by IL1B, MAP3K14, MAP3K1, IKBKB and TNF. Required for proteolytic processing of GLI3.
Specifications
| BTRC | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q9Y297 | |
| BTRC | |
| KLH conjugated synthetic peptide between 127-156 amino acids from the N-terminal region of human BRTC1/2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| BTRC | |
| BTRC;BTRCP; FBW1A; FBXW1A; F-box/WD repeat-containing protein 1A; F-box/WD repeat-containing protein 1A; E3RSIkappaB; F-box/WD repeat-containing protein 1A; Epididymis tissue protein Li 2a; F-box/WD repeat-containing protein 1A; F-box and WD repeats prote | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8945 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction