Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BubR1 Mouse anti-Human, Clone: 8G1, MilliporeSigma™

Catalog No. MAB3612MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3612MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3612MI Supplier MilliporeSigma Supplier No. MAB3612
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect BubR1 with Anti-BubR1 Antibody, clone 8G1 (Mouse Monoclonal Antibody), that has been shown to work in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BubR1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 8G1
Conjugate Unconjugated
Formulation Liquid
Gene Accession No. NM_001211.4
Gene Symbols BUB1B, Bub1A, hBUBR1, BUB1beta, SSK1, BUBR1, MAD3L, EC 2.7.11.1
Host Species Mouse
Immunogen Recombinant human BubR1.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.