Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUBR1 Mouse anti-Human, Unlabeled, Clone: 9, BD

Catalog No. BDB612502 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612502 50 μg
BDB612503 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612502 Supplier BD Biosciences Supplier No. 612502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Accurate chromosome segregation requires that all pairs of sister chromatids become appropriately atttached to mitotic spindles before the onset of anaphase. Cell cycle checkpoints monitor kinetochore-microtubule interactions, so that cell cycle progression can be delayed until proper chromosome attachments are formed. In yeast, Bub1-3 genes are required for proper mitotic delay in response to unattached kinetochores. In mammals, the homologues to yeast Bub1 and Bub3 form a complex that binds kinetochores and has protein kinase activity. BUBR1 is a bub-related kinase homologous to Bub1 and MAD3. BUBR1 contains a nuclear localization signal and two highly conserved domains CD1 and CD2. CD1 is in the N-terminal region and is invoved in kinetochore localization, while CD2 is in the C-terminal region and is involved in kinase activity. BUBR1 is hyperphosphorylated during spindle checkpoint activation, and interacts with p55CDC and APC during spindle checkpoint function. In colorectal cancers, the BUBR1 gene has structural abnormalities that suggest BUBR1 is a tumor suppressor gene. Thus, BUBR1 may be a critical spindle checkpoint kinase and disruption of its function may be related to tumor formation.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen BUBR1
Applications Western Blot
Classification Monoclonal
Clone 9
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human BUBR1 aa. 276-388
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.