Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bystin-like (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004994 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-994 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-994 Supplier Abnova Corporation Supplier No. H00000705B02

Mouse polyclonal antibody raised against a full-length human BYSL protein.

Bystin is expressed as a 2kb major transcript and a 3.6kb minor transcript in SNG-M cells and in human trophoblastic teratocarcinoma HT-H cells. Protein binding assays determined that bystin binds directly to trophinin and tastin, and that binding is enhanced when cytokeratins 8 and 18 are present. Immunocytochemistry of HT-H cells showed that bystin colocalizes with trophinin, tastin, and the cytokeratins, suggesting that these molecules form a complex in trophectoderm cells at the time of implantation. Using immunohistochemistry it was determined that trophinin and bystin are found in the placenta from the sixth week of pregnancy. Both proteins were localized in the cytoplasm of the syncytiotrophoblast in the chorionic villi and in endometrial decidual cells at the uteroplacental interface. After week 10, the levels of trophinin, tastin, and bystin decreased and then disappeared from placental villi. [provided by RefSeq

Sequence: MPKFKAARGVGGQEKHAPLADQILAGNAVRAGVREKRRGRGTGEAEEEYVGPRLSRRILQQARQQQEELEAEHGTGDKPAAPRERTTRLGPRMPQDGSDDEDEEWPTLEKAATMTAAGHHAEVVVDPEDERAIEMFMNKNPPARRTLADIIMEKLTEKQTEVETVMSEVSGFPMPQLDPRVLEVYRGVREVLSKYRSGKLPKAFKIIPALSNWEQILYVTEPEAWTAAAMYQATRIFASNLKERMAQRFYNLVLLPRVRDDVAEYKRLNFHLYMALKKALFKPGAWFKGILIPLCESGTCTLREAIIVGSIITKCSIPVLHSSAAMLKIAEMEYSGANSIFLRLLLDKKYALPYRVLDALVFHFLGFRTEKRELPVLWHQCLLTLVQRYKADLATDQKEALLELLRLQPHPQLSPEIRRELQSAVPRDVEDVPITVE

Specifications

Antigen bystin-like
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human BYSL protein.
Formulation No additive
Gene BYSL
Gene Accession No. NM_004053.3
Gene Alias BYSTIN
Gene Symbols BYSL
Host Species Mouse
Immunogen BYSL (NP_004044.3, 1 a.a. ∼ 437 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 705
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.