Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-c-Abl Clone: 19-110,

Catalog No. OP19100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP19100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP19100UG Supplier MilliporeSigma™ Supplier No. OP19100UG

Mouse Monoclonal Antibody

Specifically detects c-Abl Clone: 19-110 in Human, Mouse samples, and it is validated for Immunoprecipitation, Neutralization

The abl oncogene was initially identified as the viral transforming gene (v-abl) of Abelson murine leukemia virus (A-MuLV), from a chemically thymectomized mouse inoculated with Moloney murine leukemia virus (M-MuLV). The v-abl genome consists of M-MuLV gag sequences fused to mouse c-abl, resulting in the production of gag/c-abl tyrosine kinases, phosphoproteins p160 and p120, as the major translational products. The abl oncogene is implicated in several human leukemias including 90-95% of chronic myelocytic (CML), 20-25% of adult acute lymphoblastic (ALL) and 2-5% of pediatric ALL. In these leukemias the c-abl proto-oncogene undergoes a chromosomal translocation producing the Philadelphia (Ph1) chromosome resulting in the fusion of c-abl sequences to the bcr gene of chromosome 22. The molecular consequences of this translocation are the generation of a chimeric bcr/c-abl mRNA encoding activated abl protein-tyrosine kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Abl
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 19-110
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen a TrpE-v-Abl fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽145kDa human c-Abl, the ∽150kDa mouse c-Abl, v-Abl proteins, and the Bcr/Abl translocation products in ALL (∽190kDa) and CML (∽210kDa). Antibody Target Gene Symbol: ABL1 Target Synonym: ABL, AI325092, bcr/abl, C-ABL, C-ABL 1B, CABL1, E430008G22Rik, JTK7, MGC117749, p145Abl, p150, v-abl Entrez Gene Name: c-abl oncogene 1, receptor tyrosine kinase
Target Species Human, Mouse
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.