Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-abl Tyrosine Kinase Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3898MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3898MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3898MI Supplier MilliporeSigma Supplier No. AB3898

Rabbit Polyclonal Antibody

Detect c-abl Tyrosine Kinase using this Anti-c-abl Tyrosine Kinase Antibody, cytoplasmic and nuclear validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-abl Tyrosine Kinase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A purified immunoglobulin. Liquid in 0.02M phosphate buffer with 0.15M NaCl, pH 7.6 and 0.1% sodium azide as a preservative.
Gene Symbols ABL1; JTK7; p150; bcr/abl; v-abl; c-ABL; ABL; EC 2.7.10.2
Host Species Rabbit
Immunogen Recombinant human GST-Abl fusion protein.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005157.3; NM_007313.2
Target Species Human
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.