Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-ErbB-2 Mouse anti-Human, Unlabeled, Clone: 3B5, BD

Catalog No. BDB554299 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554299 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554299 Supplier BD Biosciences Supplier No. 554299
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C-erbB-2 (also known as HER2/neu), a 185kDa transmembrane glycoprotein, is a member of the type 1 growth factor receptor subfamily which also includes c-erbB-3, c-erbB-4 and the epidermal growth factor receptor (EGFR), also known as c-erbB-1. Members of this receptor subfamily mediate the proliferation and differentiation of normal cells. They have a common structure consisting of an extracellular domain, a transmembrane region, and a cytoplasmic sequence. The extracellular regions contain two cysteine-rich domains, and the intracellular regions have sequence homology to known tyrosine kinases. C-erbB-2 reactivity has been detected in proximal kidney tubules, mucosal epithelium in the gastrointestinal tract, and squamous epithelium in skin. Most normal adult tissues show little or no reactivity with antibodies against c-erbB-2. However, c-erbB-2 is overexpressed in many human breast, stomach, ovary and bladder carcinomas. EGFR, c-erbB-3 and c-erbB-4 are also overexpressed in various human tumor cells, and it is thought that aberrant activation of type 1 growth factor kinase activities may contribute to tumor progression.
The antibody 3B5 recognizes human c-erbB-2. A synthetic peptide corresponding to amino acids 1242-1255 (TAENPEYLGLDVPV) in the C-terminal domain of human c-erbB-2 was used as immunogen.

Host Species: Mouse
Clone: 3B5
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human c-erbB-2 aa. 1242-1255

Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-ErbB-2
Applications Western Blot
Classification Monoclonal
Clone 3B5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human c-erbB-2 aa. 1242-1255
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.