Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-erbB-3 Mouse anti-Human, Unlabeled, Clone: RTJ.1, BD

Catalog No. BDB554208 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554208 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554208 Supplier BD Biosciences Supplier No. 554208
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C-erbB-3, a glycoprotein of 160kDa, is a member of the type 1 growth factor receptor subfamily which also includes c-erbB-2 (HER2/neu), c-erbB-4 and the epidermal growth factor receptor (EGFR, c-erbB-1). Members of this receptor subfamily mediate the proliferation and differentiation of normal cells. They have a common structure consisting of an extracellular domain, a transmembrane region, and a cytoplasmic sequence. The extracellular regions contain two cysteine-rich domains, and the intracellular regions have sequence homology to known tyrosine kinases. C-erbB-3 is expressed in tissues from the digestive, urinary and respiratory tracts, the circulatory system, and female and male reproductive organs. c-erb B-3 is undetectable in hematopoietic tissue and cell lines derived from hematopoietic tumors. Cellular localization has been described as cytoplasmic and/or membrane, and nuclear. The level and pattern of c-erbB-3 expression varies widely in both normal and tumor tissues.
Clone RTJ.1 recognizes an epitope in the cytoplasmic domain of the human c-erbB-3 protein. It does not react with the EGF receptor or c-erbB-2. A synthetic peptide (referred to as 49.3) from the cytoplasmic domain of human c-erbB-3 protein was used as immunogen. RTJ.1 identifies a 160kDa band corresponding to c-erbB-3 by western blot analysis and immunoprecipitation. RTJ.1 may also react with two additional, unidentified higher molecular weight bands by immunoprecipitation and western blot analysis. These bands are likely to be non-specific, as they were not detected with a polyclonal antibody raised against the same immunogen. Because two additional bands were observed, the specificity of RTJ.1 for immunohistochemistry was analyzed by comparing staining results to those obtained using three polyclonal antibodies also raised against 49.3. Identical staining results were obtained using RTJ.1 and all three polyclonal antibodies. These results validated the use of RTJ.1 for immunohistochemical analysis of c-erbB-3.

Host Species: Mouse
Clone: RTJ.1
Isotype: IgM
Species Reactivity: Human
Immunogen: Human c-erbB-3 Peptide
Formula Weight [Chemical]: 160kDa

Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-erbB-3
Applications Immunohistochemistry
Classification Monoclonal
Clone RTJ.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human c-erbB-3 Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.