Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-erbB-3 Mouse anti-Human, Clone: RTJ2, MilliporeSigma™

Catalog No. MAB4023MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4023MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4023MI Supplier MilliporeSigma Supplier No. MAB4023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect c-erbB-3 using this Anti-c-erbB-3 Antibody, cytoplasmic domain, clone RTJ2 validated for use in ELISA, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-erbB-3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RTJ2
Conjugate Unconjugated
Formulation Liquid in PBS, containing azide as a preservative.
Gene Accession No. NM_001005915.1; NM_001982.2
Gene Symbols ERBB3, c-erbB-3, LCCS2, c-erbB3, p85-sErbB3, p45-sErbB3, ErbB-3, erbB3-S, erbB-3, MGC88033, HER3, p180-ErbB3, MDA-BF-1, EC 2.7.10.1
Host Species Mouse
Immunogen Synthetic peptide corresponding to a site on the cytoplasmic domain of the human c-erbB-3 oncoprotein
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.