Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-c-ErbB2/c-Neu Clone: 9G6,

Catalog No. OP14L100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP14L100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP14L100UG Supplier MilliporeSigma™ Supplier No. OP14L100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects c-ErbB2/c-Neu Clone: 9G6 in Human samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation

Amplification of the human proto-oncogene c-neu occurs in approximately 30% of breast carcinomas and the amplification maybe a useful predictor of disease prognosis. Gene amplification and the resulting overexpression of proto-oncogene encoded proteins is thought to transform cells by chronically stimulating signal transduction pathways, and in model systems overexpression of human c-neu induces cellular transformation. In the rat model system, a point mutation has been located within the transmembrane domain of the c-neu oncogene. neu (from neuroglioblastomas) is also referred to as erbB-2 (from erythroblastosis virus) and HER2 (for Human Epidermal growth factor Receptor) and is the second member of the EGF receptor family to be described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-ErbB2/c-Neu
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 9G6
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen cells overexpressing human c-ErbB2/c-Neu
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽185kDa c-ErbB2/c-Neu protein in SK-BR-3 cells and breast carcinoma tissue. Does not cross-react with v-ErbB2 or the EGF receptor.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.