Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-ErbB2/c-Neu Mouse anti-Human, Clone: 9G6, MilliporeSigma™

Catalog No. MABE332MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE332MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE332MI Supplier MilliporeSigma Supplier No. MABE332

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Use Anti-c-ErbB2/c-Neu Antibody, clone 9G6 (Mouse Monoclonal Antibody) validated in immunoprecipitation and immunohistochemistry to detect c-ErbB2/c-Neu also known as MLN 19, Proto-oncogene Neu or Proto-oncogene c-ErbB-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen c-ErbB2/c-Neu
Applications Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 9G6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001005862
Gene Symbols ERBB2, HER2, MLN19, NEU, NGL
Host Species Mouse
Immunogen Whole cells overexpressing human c-ErbB2/c-Neu.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.